Anti-IgA Goat Polyclonal Antibody (SureLight® APC (Allophycocyanin))

Lieferant: Abcam
ABCAAB99897-100EA 619 EUR
ABCAAB99897-100
Anti-IgA Goat Polyclonal Antibody (SureLight® APC (Allophycocyanin))
Antikörper
Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: SureLight® Allophycocyanin
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR