Anti-CD40LG Hamster Monoclonal Antibody [clone: MR-1]

Lieferant: Abcam
ABCAAB99894-100EA 872 EUR
ABCAAB99894-100
Anti-CD40LG Hamster Monoclonal Antibody [clone: MR-1]
Antikörper
Type: Primary
Antigen: CD40LG (CD40 ligand)
Clonality: monoclonal
Clone: MR-1
Conjugation: unconjugated
Epitope:
Host: Hamster
Isotype: IgG1
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR