Anti-MSH6 Rabbit monoclonal antibody [clone: SP93]

Lieferant: Abcam
ABCAAB99889-100EA 899 EUR
ABCAAB99889-100
Anti-MSH6 Rabbit monoclonal antibody [clone: SP93]
Type: Primary
Antigen: MSH6 (mutS homolog 6)
Clonality: monoclonal
Clone: SP93
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR