Anti-TNS4 Rabbit Monoclonal Antibody [clone: SP83]
Lieferant: Abcam
ABCAAB99887-100EA 847 EUR
ABCAAB99887-100
Anti-TNS4 Rabbit Monoclonal Antibody [clone: SP83]
Antikörper
Type: Primary
Antigen: TNS4 (tensin 4)
Clonality: monoclonal
Clone: SP83
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen