Anti-NLRC4 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB99860-100EA 783 EUR
ABCAAB99860-100
Anti-NLRC4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NLRC4 (NLR family, CARD domain containing 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR