Anti-IgM Mouse Monoclonal Antibody [clone: UHB] (Spectral Red®)

Lieferant: Abcam
ABCAAB99748-100EA 736 EUR
ABCAAB99748-100
Anti-IgM Mouse Monoclonal Antibody [clone: UHB] (Spectral Red®)
Antikörper
Type: Secondary
Antigen: IgM
Clonality: Monoclonal
Clone: UHB
Conjugation: Spectral Red®
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR