Anti-IgG1 Rat Monoclonal Antibody [clone: SB77e] (AP (Alkaline Phosphatase))

Lieferant: Abcam
ABCAAB99602-500EA 619 EUR
ABCAAB99602-500
Anti-IgG1 Rat Monoclonal Antibody [clone: SB77e] (AP (Alkaline Phosphatase))
Antikörper
Type: Secondary
Antigen: IgG1 (Mouse) (mouse IgG1)
Clonality: monoclonal
Clone: SB77e
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR