Anti-IgE Rat Monoclonal Antibody [clone: 23G3] (HRP (Horseradish Peroxidase))

Lieferant: Abcam
ABCAAB99574-500EA 619 EUR
ABCAAB99574-500
Anti-IgE Rat Monoclonal Antibody [clone: 23G3] (HRP (Horseradish Peroxidase))
Antikörper
Type: Secondary
Antigen: IgE (Mouse) (mouse IgE)
Clonality: monoclonal
Clone: 23G3
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR