Anti-TGFBI Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB99562-100EA 840 EUR
ABCAAB99562-100
Anti-TGFBI Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TGFBI (transforming growth factor, beta-induced, 68kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR