Anti-RPS6KA5 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB99412-100EA 859 EUR
ABCAAB99412-100
Anti-RPS6KA5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RPS6KA5 (ribosomal protein S6 kinase, 90kDa, polypeptide 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR