Anti-RTF1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB99362-100EA 859 EUR
ABCAAB99362-100
Anti-RTF1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RTF1 (Rtf1, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR