Anti-TOM1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB99356-100EA 881 EUR
ABCAAB99356-100
Anti-TOM1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TOM1 (target of myb1 (chicken))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR