Anti-ERCC3 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB99322-100EA 840 EUR
ABCAAB99322-100
Anti-ERCC3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ERCC3 (excision repair cross-complementation group 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR