Anti-PSMD4 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB98843-100EA 797 EUR
ABCAAB98843-100
Anti-PSMD4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PSMD4 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR