Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Abcam
ABCAAB98734-500EA 393 EUR
ABCAAB98734-500
Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR