Anti-IgG Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB98733-500EA 341 EUR
ABCAAB98733-500
Anti-IgG Goat Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR