Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Abcam
ABCAAB98604-500EA 393 EUR
ABCAAB98604-500
Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR