Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Abcam
ABCAAB98525-500EA 393 EUR
ABCAAB98525-500
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen