Anti-IgM Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Abcam
ABCAAB98366-500EA 393 EUR
ABCAAB98366-500
Anti-IgM Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Type: Secondary
Antigen: IgM (Rat) (rat IgM)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR