Anti-STAT5A Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB98338-100EA 823 EUR
ABCAAB98338-100
Anti-STAT5A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STAT5A (signal transducer and activator of transcription 5A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR