Anti-GOLPH3 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB98023-100EA 874 EUR
ABCAAB98023-100
Anti-GOLPH3 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: GOLPH3 (golgi phosphoprotein 3 (coat-protein))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR