Anti-HSD17B4 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB97971-50EA 840 EUR
ABCAAB97971-50
Anti-HSD17B4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HSD17B4 (hydroxysteroid (17-beta) dehydrogenase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR