Anti-PI 3 Kinase p55 gamma Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB97862-50EA 762 EUR
ABCAAB97862-50
Anti-PI 3 Kinase p55 gamma Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PI 3 Kinase p55 gamma
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR