Anti-PGAM2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB97800-50EA 840 EUR
ABCAAB97800-50
Anti-PGAM2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PGAM2 (phosphoglycerate mutase 2 (muscle))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR