Anti-GAD1 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB97739-100EA 865 EUR
ABCAAB97739-100
Anti-GAD1 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: GAD1 (glutamate decarboxylase 1 (brain, 67kDa))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR