Anti-RELA Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB97726-100EA 862 EUR
ABCAAB97726-100
Anti-RELA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RELA (v-rel avian reticuloendotheliosis viral oncogene homolog A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR