Anti-SLC5A1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB97682-100EA 841 EUR
ABCAAB97682-100
Anti-SLC5A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLC5A1 (solute carrier family 5 (sodium/glucose cotransporter), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR