Anti-ADORA2A Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB97582-50EA 840 EUR
ABCAAB97582-50
Anti-ADORA2A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADORA2A (adenosine A2a receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR