Anti-DDB1 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB97522-50EA 879 EUR
ABCAAB97522-50
Anti-DDB1 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: DDB1 (damage-specific DNA binding protein 1, 127kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR