Anti-NOVA1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB97368-50EA 840 EUR
ABCAAB97368-50
Anti-NOVA1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NOVA1 (neuro-oncological ventral antigen 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR