Anti-MAGI2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB97343-50EA 762 EUR
ABCAAB97343-50
Anti-MAGI2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAGI2 (membrane associated guanylate kinase, WW and PDZ domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR