Anti-TULP1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB97281-50EA 797 EUR
ABCAAB97281-50
Anti-TULP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TULP1 (tubby like protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR