Anti-IgG3 Goat Polyclonal Antibody (Biotin)

Lieferant: Abcam
ABCAAB97258-1EA 393 EUR
ABCAAB97258-1
Anti-IgG3 Goat Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: IgG3 (Mouse) (mouse IgG3)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR