Anti-IgG2b Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Lieferant: Abcam
ABCAAB97249-1EA 393 EUR
ABCAAB97249-1
Anti-IgG2b Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Type: Secondary
Antigen: IgG2b (Mouse) (mouse IgG2b)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen