Anti-IgG1 Goat Polyclonal Antibody (Biotin)
Lieferant: Abcam
ABCAAB97238-1EA 393 EUR
ABCAAB97238-1
Anti-IgG1 Goat Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: IgG1 (Mouse) (mouse IgG1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen