Anti-IgA Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Abcam
ABCAAB97234-1EA 393 EUR
ABCAAB97234-1
Anti-IgA Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Type: Secondary
Antigen: Mouse IgA
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR