Anti-IgA Goat Polyclonal Antibody (Biotin)
Lieferant: Abcam
ABCAAB97183-1EA 393 EUR
ABCAAB97183-1
Anti-IgA Goat Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: Rat IgA
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen