Anti-IgA Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB97181-1EA 324 EUR
ABCAAB97181-1
Anti-IgA Goat Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: Rat IgA
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR