Anti-IgG Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB97136-1EA 359 EUR
ABCAAB97136-1
Anti-IgG Rabbit Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR