Anti-IgG Donkey Polyclonal Antibody (Biotin)

Lieferant: Abcam
ABCAAB97123-1EA 393 EUR
ABCAAB97123-1
Anti-IgG Donkey Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: IgG (Sheep) (sheep IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Donkey
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR