Anti-IgM Goat Polyclonal Antibody (DyLight® 650)

Lieferant: Abcam
ABCAAB96982-500EA 377 EUR
ABCAAB96982-500
Anti-IgM Goat Polyclonal Antibody (DyLight® 650)
Antikörper
Type: Secondary
Antigen: Rabbit IgM
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR