Anti-IgG Goat Polyclonal Antibody (DyLight® 550)

Lieferant: Abcam
ABCAAB96948-500EA 393 EUR
ABCAAB96948-500
Anti-IgG Goat Polyclonal Antibody (DyLight® 550)
Antikörper
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR