Anti-IgG Donkey Polyclonal Antibody (DyLight® 488)

Lieferant: Abcam
ABCAAB96939-500EA 393 EUR
ABCAAB96939-500
Anti-IgG Donkey Polyclonal Antibody (DyLight® 488)
Antikörper
Type: Secondary
Antigen: IgG (Sheep) (sheep IgG)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Donkey
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR