Anti-IgG Donkey Polyclonal Antibody (DyLight® 650)
Lieferant: Abcam
ABCAAB96934-500EA 393 EUR
ABCAAB96934-500
Anti-IgG Donkey Polyclonal Antibody (DyLight® 650)
Antikörper
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Donkey
Isotype: IgG
Reactivity:
Bestellen