Anti-IgG Rabbit Polyclonal Antibody (DyLight® 650)

Lieferant: Abcam
ABCAAB96918-500EA 448 EUR
ABCAAB96918-500
Anti-IgG Rabbit Polyclonal Antibody (DyLight® 650)
Antikörper
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR