Anti-GNRPX Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB96822-50EA 797 EUR
ABCAAB96822-50
Anti-GNRPX Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: GNRPX
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR