Anti-FIS1 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB96764-50EA 862 EUR
ABCAAB96764-50
Anti-FIS1 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: FIS1 (fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR