Anti-CD55 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB96680-50EA 840 EUR
ABCAAB96680-50
Anti-CD55 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CD55 (CD55 molecule, decay accelerating factor for complement (Cromer blood group))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR