Anti-CFL2 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB96678-50EA 859 EUR
ABCAAB96678-50
Anti-CFL2 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: CFL2 (cofilin 2 (muscle))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR