Anti-RBM4 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB96658-50EA 763 EUR
ABCAAB96658-50
Anti-RBM4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RBM4 (RNA binding motif protein 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR